missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-PRAS40 (T246) DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC6890-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444775

  • £303.61 / 1 set

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
84335
2 Plates
Human
Videos