missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-PDGF R alpha DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC2114-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464435

  • £349.81 / Pack of 2


Estimated Shipment
28-02-2019
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
5156
2 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Videos