missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-p53 (S15) DuoSet IC, R&D Systems™

Human Phospho-p53 (S15) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC1839E

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434405

  • £2146.20 / Pack of 15

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human Phospho-p53 (S15) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human p53.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
7157
15 Plates
Human
312.00 - 20000pg/mL
Videos
Documents