missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-MSP R/Ron DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
Brand: R&D Systems™ DYC1947-2
Code : Z1
Additional Details : Weight : 0.01000kg
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits
Specifications
ELISA | |
4486 | |
2 Plates | |
Human |
Biotin | |
ELISA Development Kit for phosphorylated intracellular proteins | |
1 Kit |