missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-MKK3 (S218/T222) DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC5585-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464715

  • £303.61 / Pack of 2


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
5606
2 Plates
Human
Videos