missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-Mer DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Gene ID (Entrez) | 10461 |
---|---|
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
The Human Phospho-Mer DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Mer.Specifications
10461 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |