Learn More
R&D Systems™ Human Phospho-Mer DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
Brand:  R&D Systems™ DYC2579-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13434505
  • £751.17 / Pack of 5
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
The Human Phospho-Mer DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Mer.
Specifications
Specifications
ELISA
10461
5 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.