missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-Lyn (Y397) DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC3936-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434635

  • £371.91 / Pack of 2

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Descripción

Descripción

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Especificaciones

Especificaciones

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
4067
2 Plates
Human
Videos
Documentos