missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-Insulin R DuoSet IC, R&D Systems™
Human Phospho-Insulin R DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Gene ID (Entrez) | 3643 |
---|---|
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
The Human Phospho-Insulin R DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Insulin R/CD220.Specifications
3643 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |