Learn More
R&D Systems™ Human Phospho-Insulin R DuoSet IC, R&D Systems™
Human Phospho-Insulin R DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC2718-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13434515
  • £751.17 / Pack of 5
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
The Human Phospho-Insulin R DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Insulin R/CD220.
Specifications
Specifications
ELISA
3643
5 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.