missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-IGF-I R DuoSet IC, R&D Systems™
Human Phospho-IGF-I R/IGF1R DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Gene ID (Entrez) | 3480 |
---|---|
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
The Human Phospho-IGF-I R/IGF1R DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human IGF-I R/IGF1R.Specifications
3480 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |