Learn More
R&D Systems™ Human Phospho-HGFR/c-MET DuoSet IC ELISA
Human Phospho-HGF R/c-MET DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC2480E
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13434485
  • £2210.88 / Pack of 15
Estimated Shipment date
from Supplier 02-04-2020
Add to basket
Description
Description
The Human Phospho-HGF R/c-MET DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human HGFR/c-MET.
Specifications
Specifications
ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Human
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
1 Kit
4233
15 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.