missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-HGFR/c-MET DuoSet IC ELISA
Human Phospho-HGF R/c-MET DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Gene ID (Entrez) | 4233 |
---|---|
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
The Human Phospho-HGF R/c-MET DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human HGFR/c-MET.Specifications
4233 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |