missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-Flt-3/Flk-2 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

POA

Specifications

Gene ID (Entrez) 2322
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Conjugate Biotin
Species Human
View More Specs

Description

Description

The Human Phospho-Flt-3/Flk-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Flt-3/Flk-2/CD135.
Specifications

Specifications

2322
ELISA Development Kit for phosphorylated intracellular proteins
Biotin
Human