Learn More
R&D Systems™ Human Phospho-Flt-3/Flk-2 DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
Brand:  R&D Systems™ DYC368E
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13444605
  • £2146.20 / Pack of 15
Call for latest availability
+44 (0)1509 555 500
Description
Description
The Human Phospho-Flt-3/Flk-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Flt-3/Flk-2/CD135.
Specifications
Specifications
ELISA
2322
15 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.