Learn More
R&D Systems™ Human Phospho-FGFR3 DuoSet IC ELISA
Human Phospho-FGF R3 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC2719-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13454515
  • £852.60 / Pack of 5
Estimated Shipment date
from Supplier 02-04-2020
Add to basket
Description
Description
The Human Phospho-FGF R3 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human FGFR3.
Specifications
Specifications
ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Human
CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
1 Kit
2261
5 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.