Learn More
R&D Systems™ Human Phospho-FGF R3 DuoSet IC, R&D Systems™
Human Phospho-FGF R3 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC2719-2
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13444515
  • £384.13 / Pack of 2
Estimated Shipment
09-07-2019
Description
Description
The Human Phospho-FGF R3 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human FGFR3.
Specifications
Specifications
ELISA
2261
2 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos