missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-FGFR2 alpha DuoSet IC ELISA

Phospho-FGF R2 alpha DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC684-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454765

  • £401.31 / Pack of 2


Estimated Shipment date
from Supplier 03-04-2020
Add to basket

Description

Description

The Phospho-FGF R2 alpha DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human FGFR2 alpha.
Specifications

Specifications

ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Human
1 Kit
2263
2 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
Documents