missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-ErbB3/Her3 DuoSet IC, R&D Systems™

Human Phospho-ErbB3/Her3 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC1769E

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454385

  • £2210.88 / Pack of 15


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Human Phospho-ErbB3/Her3 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human ErbB3/Her3.
Specifications

Specifications

ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Human
1 Kit
2065
15 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
Documents