missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-ErbB2/Her2 DuoSet IC, R&D Systems™

Human Phospho-ErbB2/Her2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

POA

Specifications

Gene ID (Entrez) 2064
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Conjugate Biotin
Species Human
View More Specs

Description

Description

The Human Phospho-ErbB2/Her2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human ErbB2/Her2.
Specifications

Specifications

2064
ELISA Development Kit for phosphorylated intracellular proteins
Biotin
Human
Videos
SDS
Documents

Documents

Product Certifications