missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-EphA2 DuoSet IC, R&D Systems™
Human Phospho-EphA2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Gene ID (Entrez) | 1969 |
---|---|
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Species | Human |
Description
ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the simultaneous detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no ass
The Human Phospho-EphA2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human EphA2.Specifications
1969 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |