missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-EGF R/ErbB1 (Y1068) DuoSet IC, R&D Systems™
Human Phospho-EGFR (Y1068) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 312.00 to 20000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 1956 |
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Conjugate | Biotin |
Description
ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of . Complete ELISpot kits are ready-to-run and require no assay development or refinement.
The Human Phospho-EGFR (Y1068) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human EGFR.Specifications
312.00 to 20000 pg/mL | |
1956 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Human |