missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-DDR1 DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
---|---|
Species | Human |
Description
The Human Phospho-DDR1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human DDR1.Specifications
ELISA Development Kit for phosphorylated intracellular proteins | |
Human |