Learn More
Invitrogen™ PEI-Transferrinfection Kit
Nucleic acid delivery system based on transferrin receptor-mediated endocytosis to carry DNA into cells and the gene transfer efficiency of PEI/DNA complexes
Brand: Invitrogen™ BMS1003
Code : NEW
Additional Details : Weight : 0.47500kg
Includes: Transferrin-Polyethylenimine-conjugate, lyophilized, 1 mg/mL upon reconstitution; iron incorporated
Deferrioxamine, lyophilied, 50 mM upon reconstitution
HBS buffer concentrate (20x)
(400mM Hepes, pH 7.3, 3M NaCl)
Description
- Method results in a 30 to 1000-fold enhanced transfection efficiency depending on the cell line
- An extremely gentle DNA transfection method which employs physiological uptake mechanisms of the cell
- Transfection efficacy depends on the cell type, the level of surface transferrin receptor expression
- Very high transfection rates have been shown for the tested tumor cell lines B16F10 melanoma, Neuro 2A neuroblastoma and a variety of primary human melanoma cell lines
- In other established cell lines, such as HeLa, CHO, Jurkat, K562, HepG2 and COS the PEI-Transferrinfection works with high efficiences, excellent reproducibility and with the advantage of being an extremely gentle procedure
Specifications
Chemically Defined | |
96-well Plate | |
Transferrin Receptor-mediated Endocytosis | |
Transferrinfection Kit |
Tumor Cell Lines, Established Cell Lines | |
Plasmid DNA | |
Transfection | |
32 Transferrinfections |