Learn More
Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
£96.55 - £580.00
Specifications
Antigen | Human Papilloma Virus (HPV) Ab-3 |
---|---|
Clone | K1H8 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12620227
|
Epredia™
MS-1826-S0 |
100 μL |
£96.55
100µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12630227
|
Epredia™
MS-1826-S1 |
500 μL |
£580.00
500µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12610227
|
Epredia™
MS-1826-S |
1 mL |
£480.00
1mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12690217
|
Epredia™
MS-1826-R7 |
7 mL |
£209.00
7mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12600227
|
Epredia™
MS-1826-RQ |
12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.Host Species: Mouse
Clone: K1H8
Isotype: IgG1/κ
Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.
Epitope: Nonconfirmational internal linear epitope of major capsid protein
Immunogen: Alkaline disrupted HPV type 1
Molecular Weight: 57kDa (major band)
Positive Control: Infected cells and tissue
Cellular Localization: Nuclei of infected cells
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Human Papilloma Virus (HPV) Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
ASR, IVD | |
Alkaline disrupted HPV type 1 |
K1H8 | |
Monoclonal | |
Mouse | |
Infectious Disease | |
Primary |