Learn More
Abnova™ Human OTP Partial ORF (NP_115485.1, 11 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023440-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development. [provided by RefSeq]
Sequence: RLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELESpecifications
NP_115485.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELE | |
RUO | |
OTP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23440 | |
OTP (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3161 | |
OTP | |
Recombinant | |
wheat germ expression system |