Learn More
Invitrogen™ Osteopontin Human ELISA Kit
For the quantitative detection of human Osteopontin.
Brand: Invitrogen™ BMS2066TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human Osteopontin
Biotin-Conjugate anti-human Osteopontin polyclonal antibody
Streptavidin-HRP
Human Osteopontin Standard lyophilized, 60ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Osteopontin (Hu OPN) ELISA quantitates Hu OPN in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu OPN. Principle of the method The Human OPN solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in malignant glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.Specifications
P10451 | |
0.26 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
BNSP,BSPI,ETA-1,OPN | |
6.7% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 30 min. |
0.47-30 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
6696 | |
6.1% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
OPN, ETA-1, BSP-1, SPP1 | |
1 hr. 20 min. |
For Research Use Only