missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Osteopontin (OPN) Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human Osteopontin in cell culture supernates, plasma, urine, and human milk.

Brand:  R&D Systems™ DOST00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462705

  • £539.52 / 96 tests


Estimated Shipment
28-02-2019
Add to basket

Description

Description

It contains NS0-expressed recombinant human Osteopontin and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human Osteopontin showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring human Osteopontin
Specifications

Specifications

0.31 - 20ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
6696
1 Plate
0.024ng/mL
Cell Culture Supernates (50μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Urine (50μL), Human Milk (50μL)
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Urine (50uL), Human Milk (50uL)
Human
Videos