missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NR1I3 Partial ORF (NP_005113.1, 2 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009970-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005205

  • £275.34 / 10µg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: ASREDELRNCVVCGDQATGYHFNALTCEGCKGFFRRTVSKSIGPTCPFAGSCEVSKTQ
Specifications

Specifications

NP_005113.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.12kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASREDELRNCVVCGDQATGYHFNALTCEGCKGFFRRTVSKSIGPTCPFAGSCEVSKTQ
RUO
NR1I3
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9970
NR1I3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAR|CAR1|MB67|MGC150433|MGC97144|MGC97209
NR1I3
Yes
Videos
Documents