missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NR1I3 Partial ORF (NP_005113.1, 2 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

£280.00 - £425.00

Specifications

Accession Number NP_005113.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9970
Molecular Weight (g/mol) 32.12kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16005205
View Documents Promotion Details
Abnova™
H00009970-Q01.10UG
10 ug
£280.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16015205
View Documents Promotion Details
Abnova™
H00009970-Q01.25UG
25 ug
£425.00
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists. Ligand binding results in translocation of this protein to the nucleus, where it activates or represses target gene transcription. These ligands include bilirubin, a variety of foreign compounds, steroid hormones, and prescription drugs. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ASREDELRNCVVCGDQATGYHFNALTCEGCKGFFRRTVSKSIGPTCPFAGSCEVSKTQ
Specifications

Specifications

NP_005113.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.12kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAR/CAR1/MB67/MGC150433/MGC97144/MGC97209
NR1I3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9970
NR1I3 (Human) Recombinant Protein (Q01)
ASREDELRNCVVCGDQATGYHFNALTCEGCKGFFRRTVSKSIGPTCPFAGSCEVSKTQ
RUO
NR1I3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications