Learn More
R&D Systems™ Human/Mouse Total COX-2 DuoSet IC, R&D Systems™
Human/Mouse Total COX-2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Product Type | ELISA Development Kit for intracellular proteins |
Species | Human, Mouse |
Description
The Human/Mouse Total COX-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse COX-2.Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human, Mouse |