Learn More
Invitrogen™ TGF beta-1 Human/Mouse Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human or mouse TGF beta 1 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range of 8-1000pg/mL.
£158.00 - £546.00
Specifications
Accession Number | P01137,P04202 |
---|---|
Assay Range | 8-1,000 pg/mL |
Assay Sensitivity | 8 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15521227
|
Invitrogen™
88-8350-76 |
20 x 96 Tests |
£546.00
Pack of 20 |
In Stock
Log in to see stock. |
|||||
15541227
|
Invitrogen™
88-8350-86 |
10 x 96 Tests |
£391.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15298897
|
Invitrogen™
88-8350-22 |
2 x 96 Tests |
£158.00
2 sets |
In Stock
Log in to see stock. |
|||||
Description
The Human/Mouse Transforming Growth Factor beta 1 (TGFβ1) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human/mouse TGFβ1. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human/Mouse TGF beta 1 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
P01137,P04202 | |
8 pg/mL | |
ELISA Kit | |
Human, Mouse | |
Colorimetric Microplate Reader | |
IFN-g,Ifg | |
HRP | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 100 μL | |
Transforming Growth Factor beta 1, TGF-b1 | |
1 hr. 45 min. |
8-1,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
21803, 7040 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards., 96-well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22; -76; and -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
For Research Use Only