missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human/Mouse TGF beta 1 ELISA Ready-SET-Go!™ Kit, 2nd Generation

Specifically engineered for accurate and precise measurement of human or mouse TGF beta 1 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range of 8-1000pg/mL.

Brand:  Invitrogen™ 88-8350-88

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551227

  • £280.64 / Pack of 10


EU Stock 6
Add to basket

Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.

Description

Description

Human/Mouse TGF beta 1 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
8pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Transforming Growth Factor beta 1, TGF-b1
10 x 96 tests
Human, Mouse
TGF beta 1
Videos

For Research Use Only