missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Total TC-PTP DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC1930-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464415

  • £303.61 / Pack of 2


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
5771
1 Kit
Two 96-well plates
ELISA Development Kit for intracellular proteins
Human, Mouse, Rat
Videos