missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Total SOD2/Mn-SOD DuoSet IC, R&D Systems™

Human/Mouse/RatTotal SOD2/Mn-SOD DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC3419-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444585

  • £349.81 / Pack of 2

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human/Mouse/RatTotal SOD2/Mn-SOD DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat SOD2/Mn-SOD.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
6648
2 Plates
Human, Mouse, Rat
Videos