missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC, R&D Systems™

Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC1663-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444355

  • £827.61 / Pack of 5

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat HSP70/HSPA1A.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
24472
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
Five 96-well plates
ELISA Development Kit for intracellular proteins
Human, Mouse, Rat
156.00 - 10000pg/mL
Videos
Documents