missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Total GSK-3 alpha/beta DuoSet IC, R&D Systems™

POA

Specifications

For Use With (Application) Development of sandwich ELISA to measure GSK-3 alpha/beta in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Kit
Quantity 1 Kit
Species Human, Mouse, Rat
View More Specs

Description

Description

FLIP antibody specifically detects FLIP in Human samples. It is validated for Proximity Ligation Assay.
Specifications

Specifications

Development of sandwich ELISA to measure GSK-3 alpha/beta in cell lysates
ELISA Kit
Human, Mouse, Rat
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit