missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Assay Range | 312.50 to 20000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 20104 |
Product Type | ELISA Development Kit for phosphorylated intracellular proteins |
Format | 15 96-well plates |
Description
The Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat Ribosomal Protein S6/RPS6.Specifications
312.50 to 20000 pg/mL | |
20104 | |
15 96-well plates | |
Human, Mouse, Rat |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin |