missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC3918-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434625

  • £401.31 / Pack of 2


Estimated Shipment date
from Supplier 26-02-2020
Add to basket

Description

Description

The Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat Ribosomal Protein S6/RPS6.
Specifications

Specifications

ELISA
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
2 96-well plates
Biotin
29304
Human, Mouse, Rat
312.50 to 20000 pg/mL
Videos
Documents