Learn More
R&D Systems™ Human/Mouse/Rat BMP-2 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid-phase ELISA designed to measure human BMP-2 in bone tissue extracts and cell culture supernates.
Brand:  R&D Systems™ SBP200
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462666
  • £3037.79 / Pack of 6
Estimated Shipment
28-02-2019
Add to basket
Description
Description
It contains CHO cell-expressed recombinant human BMP-2 and has been shown to accurately quantitate the recombinant factor. Results obtained using natural mouse BMP-2 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring human BMP-2
Specifications
Specifications
62.50 - 4000pg/mL
<50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
650 (Human); 12156 (Mouse); 29373 (Rat)
6 Plates
29pg/mL
Cell Culture Supernates (50μL), Bone Extracts (50μL)
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Bone Extracts (50uL)
Human, Mouse, Rat
Videos