missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse Cleaved Caspase-3 (Asp175) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC835-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464805

  • £319.45 / Pack of 2


Estimated Shipment
03-09-2018
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Biotin
12367
2 Plates
Human, Mouse
Videos