missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD45R / B220 Rat, Clone: RA3-6B2, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £146.00
Specifications
Antigen | Human/Mouse CD45R / B220 |
---|---|
Clone | RA3-6B2 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951978
|
EnquireBio
QAB41-100UG |
100 μg |
£71.60
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961978
|
EnquireBio
QAB41-500UG |
500 μg |
£146.00
500µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Human/Mouse CD45R / B220 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
19264 | |
IgG2a κ | |
Protein A/G Purified |
RA3-6B2 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
NP_001255215 | |
Ptprc | |
Primary | |
Store at 2-4°C. |