missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD44 Rat, PE-Cy7, Clone: IM7, enQuireBio™
Rat Monoclonal Antibody
£89.95 - £191.00
Specifications
Antigen | Human/Mouse CD44 |
---|---|
Clone | IM7 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951938
|
EnquireBio
QAB39-PE7-25UG |
25 μg |
£89.95
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941938
|
EnquireBio
QAB39-PE7-100UG |
100 μg |
£191.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Human/Mouse CD44 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1, pH7.2 | |
12505 | |
IgG2b κ | |
Protein A/G Purified |
IM7 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_001034239 | |
Cd44 | |
Primary | |
Store at 2-4°C. |