missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human MMP-9 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human MMP-9.

Brand:  Invitrogen™ BMS2016-2CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581487

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-9
Biotin-Conjugate anti-human MMP-9 polyclonal antibody
Streptavidin-HRP
Human MMP-9 Standard lyophilized, 30ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (citrate, heparin)
Human
92 kDa gelatinase, Gelatinase B, GELB
ELISA
0.05ng/mL
10μL (1:25 prediluted)
MMP-9
Videos

For Research Use Only