eBioscience Human MMP-9 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human MMP-9.

Overview
Brand: eBioscience

Manufacturer Part Number: BMS2016/2TENCE

X10 Human MMP-9 Platinum ELISA (supplied as 10 x 96 tests)

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-9
Biotin-Conjugate anti-human MMP-9 polyclonal antibody
Streptavidin-HRP
Human MMP-9 Standard lyophilized, 30ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Product Code. 15501497

Quantity Price
1 £ 3287.1 / Pack of 10
Estimated Shipment date
from Supplier 27-02-2017
Add to basket


Description and Specification

Specification

Sample Type Cell culture supernatant, serum, plasma (citrate, heparin)
Sample Size 10μL (1:25 prediluted)
Synonym 92 kDa gelatinase, Gelatinase B, GELB
For Use With (Application) ELISA
Quantity 10 x 96 tests
Sensitivity 0.05ng/mL
Species Human
Target MMP-9
Conjugate Biotin

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.