missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP3 Human ELISA Kit
For the quantitative detection of total human MMP-3.
Brand: Invitrogen™ BMS2014-3TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-3
Description
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form,and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions,and has metallo-endopeptidase activity. The active enzyme degrades collagen,gelatin,fibronectin,and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
P08254 | |
0.0005ng/mL | |
Plasma, Serum, Supernatant | |
ELISA | |
4314 | |
4% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 30 min. |
0.06-4.00ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
MMP3 | |
6.1% | |
10 x 96 Tests | |
Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL | |
Matrix metalloproteinase-3, Stromelysin-1, Progelatinase, Transin | |
1 hr. 20 min. |
For Research Use Only