Learn More
Invitrogen™ MMP10 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHMMP10
Code : NEW
Additional Details : Weight : 0.00400kg
Description
The Human Matrix MetalloProteinase 10 (Hu MMP10) ELISA quantitates Hu MMP10 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MMP10. Principle of the method The Human MMP10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.Specifications
P09238 | |
3 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
4319 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Extracellular Matrix & Cell Adhesion | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.06-1500 pg/mL | |
3pg/mL to 1500pg/mL | |
Biotin | |
Cell Structure & Organelle Proteins | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, EGF, ENA-78, FGF-4, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, 6, 7, 9, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, MMP-1, -2, -3, -9,- 12,-13, PARC, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
SL-2,STMY2 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
SL-2, STMY2 | |
1 hr. 20 min. |