missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MIF Quantikine ELISA Kit, R&D Systems™

Human MIF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DMF00B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442695

  • £610.05 / 96 tests

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human MIF Quantikine ELISA Kit quantifies human MIF in cell culture supernates (50 ul), serum (10 ul), platelet-poor edta plasma (10 ul), platelet-poor heparin plasma (10 ul), urine (10 ul), human milk (10 ul).
Specifications

Specifications

0.16 - 10ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
4282
1 Plate
0.068ng/mL
Cell Culture Supernates (50μL), Serum (10μL), Platelet-poor EDTA Plasma (10μL), Platelet-poor Heparin Plasma (10μL), Urine (10μL), Human Milk (10μL
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), Serum (10uL), Platelet-poor EDTA Plasma (10uL), Platelet-poor Heparin Plasma (10uL), Urine (10uL), Human Milk (10uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents