missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MIF Quantikine ELISA Kit, R&D Systems™
Human MIF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.16 - 10ng/mL |
---|---|
Assay Sensitivity | 0.068ng/mL |
Sample Type | Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA, Heparin), Serum, Urine |
Description
The Human MIF Quantikine ELISA Kit quantifies human MIF in cell culture supernates (50 ul), serum (10 ul), platelet-poor edta plasma (10 ul), platelet-poor heparin plasma (10 ul), urine (10 ul), human milk (10 ul).Specifications
0.16 - 10ng/mL | |
0.068ng/mL | |
Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA, Heparin), Serum, Urine |