missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human MCP-1 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human MCP-1.

Brand:  Invitrogen™ BMS281CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15541877

  • £359.99 / 96 tests


EU Stock 37
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MCP-1
HRP-Conjugate anti-human MCP-1 monoclonal antibody
Human MCP-1 Standard lyophilized, 2ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
96 tests
Cell culture supernatant, serum, plasma (EDTA, heparin)
Human
Monocyte chemoattractant protein 1, MCAF, HC11
ELISA
2.3pg/mL
20μL
CCL2 (MCP-1)
Videos

For Research Use Only